missing translation for 'onlineSavingsMsg'
Learn More

Methyl hydrogen terephthalate, 99+%

Product Code. 15440698
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15440698 5 g 5g
15450698 25 g 25g
15460698 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15440698 Supplier Thermo Scientific Alfa Aesar Supplier No. H54391.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1679-64-7 | C9H8O4 | 180.159 g/mol

Methyl hydrogen terephthalate is used in the synthesis of Hepatitis C antiviral activity. It is also used in the design and preparation of benzamide derivatives as BRAFV600E inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl hydrogen terephthalate is used in the synthesis of Hepatitis C antiviral activity. It is also used in the design and preparation of benzamide derivatives as BRAFV600E inhibitors.

Solubility
Slightly soluble in water. Also soluble in methanol, benzene and ether.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 1679-64-7
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.159
MDL Number MFCD00002557
InChI Key REIDAMBAPLIATC-UHFFFAOYSA-N
Synonym 4-methoxycarbonyl benzoic acid, mono-methyl terephthalate, monomethyl terephthalate, methyl terephthalate, methyl hydrogen terephthalate, 1,4-benzenedicarboxylic acid, monomethyl ester, terephthalic acid monomethyl ester, 1,4-benzenedicarboxylic acid monomethyl ester, terephthalic acid, monomethyl ester, hydrogen methyl terephthalate
PubChem CID 15513
IUPAC Name 4-methoxycarbonylbenzoic acid
SMILES COC(=O)C1=CC=C(C=C1)C(=O)O

Specifikationer

Melting Point 220°C to 223°C
Quantity 5 g
Beilstein 1911082
Solubility Information Slightly soluble in water. Also soluble in methanol,benzene and ether.
Formula Weight 180.16
Percent Purity ≥99%
Chemical Name or Material Methyl hydrogen terephthalate

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.