missing translation for 'onlineSavingsMsg'
Learn More

Methyl hydrogen terephthalate, 99+%

Product Code. p-7036142
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 15440698

Brand: Thermo Scientific Alfa Aesar H54391.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1679-64-7 | C9H8O4 | 180.159 g/mol

Methyl hydrogen terephthalate is used in the synthesis of Hepatitis C antiviral activity. It is also used in the design and preparation of benzamide derivatives as BRAFV600E inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl hydrogen terephthalate is used in the synthesis of Hepatitis C antiviral activity. It is also used in the design and preparation of benzamide derivatives as BRAFV600E inhibitors.

Solubility
Slightly soluble in water. Also soluble in methanol, benzene and ether.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1679-64-7
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.159
MDL Number MFCD00002557
InChI Key REIDAMBAPLIATC-UHFFFAOYSA-N
Synonym 4-methoxycarbonyl benzoic acid, mono-methyl terephthalate, monomethyl terephthalate, methyl terephthalate, methyl hydrogen terephthalate, 1,4-benzenedicarboxylic acid, monomethyl ester, terephthalic acid monomethyl ester, 1,4-benzenedicarboxylic acid monomethyl ester, terephthalic acid, monomethyl ester, hydrogen methyl terephthalate
PubChem CID 15513
IUPAC Name 4-methoxycarbonylbenzoic acid
SMILES COC(=O)C1=CC=C(C=C1)C(=O)O

Specifications

Melting Point 220°C to 223°C
Quantity 5 g
Beilstein 1911082
Solubility Information Slightly soluble in water. Also soluble in methanol,benzene and ether.
Formula Weight 180.16
Percent Purity ≥99%
Chemical Name or Material Methyl hydrogen terephthalate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.