missing translation for 'onlineSavingsMsg'
Learn More

Methyltestosterone (17alpha-Methyltestosterone) 1.0 mg/ml in Acetonitrile, LoGiCal

Product Code. 30458051
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30458051 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30458051 Supplier LoGiCal Supplier No. LGCAMP0558.0011

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 17-alpha-Methyltestosterone
CAS 58-18-4
Analyte or Component Names 17-alpha-Methyltestosterone
Shipping Condition Room Temperature
Molecular Formula C20 H30 O2
Certifications/Compliance ISO 17034
Synonym Methyltestosterone, 17alpha-Methyltestosterone, 17-Hydroxy-17-methyl-3-keto-androstene-4, 17-Methyltestosteron, 17-Methyltestosterone, Android, Andrometh, Androsan, Androsan (tablets), Androsten, Anertan, Anertan (tablets), CDB 110, Dumogran, Glosso-Sterandryl, Homandren, Hormale, L 589.372, M.T.Mucorettes, Malestrone, Malogen, Masenone, Mastestona, Mesterone, Metandren, Methyltestorone, Metrone, NSC 139965, NSC 9701, Neo-Hombreol-M, Nu-man, Oraviron, Orchisterone M, Oreton Methyl, Oreton-M, Perandren, RU 24400, Steronyl, Synandrets, Synandrotabs, Testhormone, Testora, Testoviron, Testoviron (tablet), Testovis Depot, Testred, U 2842, Androst-4-en-3-one, 17-hydroxy-17-methyl-, (17beta)-, Androst-4-en-3-one, 17beta-hydroxy-17-methyl- (8CI), (17beta)-17-Hydroxy-17-methylandrost-4-en-3-one, 17(alpha)-Methyl-Delta4-androsten-17-(beta)-ol-3-one, 17alpha-Methyl-17beta-hydroxyandrost-4-en-3-one, 17alpha-Methyl-17beta-hydroxyandrost-4-ene-3-one, 17alpha-Methyl-3-oxo-4-androsten-17beta-ol, 17alpha-Methylandrost-4-en-17-ol-3-one, 17alpha-Methylandrost-4-en-17beta-ol-3-one, 17beta-Hydroxy-17-methylandrost-4-en-3-one, 17beta-Hydroxy-17alpha-methyl-3-oxoandrost-4-ene, 17beta-Hydroxy-17alpha-methylandrost-4-en-3-one, 17beta-Hydroxy-17alpha-methylandrost-4-ene-3-one, Androst-4-ene-17alpha-methyl-17beta-ol-3-one
InChI Formula InChI=1S/C20H30O2/c1-18-9-6-14(21)12-13(18)4-5-15-16(18)7-10-19(2)17(15)8-11-20(19,3)22/h12,15-17,22H,4-11H2,1-3H3/t15-,16+,17+,18+,19+,20+/m1/s1
Solution Type Single Solution
SMILES C[C@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC Name (8R,9S,10R,13S,14S,17S)-17-hydroxy-10,13,17-trimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one
Solvent or Matrix Acetonitrile
Industry Type Clinical Testing
Molecular Weight (g/mol) 302.45
Quantity 1 mL
Formula Weight 302.2246 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.