missing translation for 'onlineSavingsMsg'
Learn More

Methylthiouracil, Mikromol™

Product Code. 17991206
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17991206 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17991206 Supplier Mikromol Supplier No. MM3148.00
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Methylthiouracil
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6-Methyl-2-thiouracil
CAS 56-04-2
Analyte or Component Names 6-Methyl-2-thiouracil
Shipping Condition Room Temperature
Molecular Formula C5 H6 N2 O S
Certifications/Compliance ISO 17034
Synonym 4(1H)-Pyrimidinone, 2,3-dihydro-6-methyl-2-thioxo-, Uracil, 6-methyl-2-thio- (8CI), 2-Mercapto-4-methyl-6-hydroxypyrimidine, 2-Mercapto-6-methyl-4-pyrimidinol, 2-Mercapto-6-methylpyrimidin-4-one, 2-Mercapto-6-methylpyrimidyl-4-one, 2-Thio-6-methyluracil, 4-Hydroxy-2-mercapto-6-methylpyrimidine, 4-Hydroxy-6-methyl-2-mercaptopyrimidine, 4-Hydroxy-6-methylpyrimidine-2(1H)-thione, 4-Methyl-2-thiouracil, 6-Methyl-2-mercaptouracil, 6-Methyl-2-thio-2,4-(1H,3H)-pyrimidinedione, 6-Methyl-2-thiouracil, 6-Methyl-4-oxo-2-thioxo-1,2,3,4-tetrahydropyrimidine, 6-Thio-4-methyluracil, Alkiron, Antibason, Basecil, Basethyrin, MTU, Metacil, Methacil, Methiacil, Methicil, Methiocil, Methylthiouracil, Metiur, Muracil, Muracin, NSC 193526, NSC 314271, NSC 9378, Orcanon, Prostrumyl, Strumacil, Thimecil, Thiomecil, Thiomidil, Thioryl, Thiothymin, Thiothyron, Thiuryl, Thyreonorm, Thyril, Tiomeracil, Tiorale M, Tiotiron, USAF-EK 6454
InChI Formula InChI=1S/C5H6N2OS/c1-3-2-4(8)7-5(9)6-3/h2H,1H3,(H2,6,7,8,9)
SMILES CC1=CC(=O)NC(=S)N1
IUPAC Name 6-methyl-2-sulfanylidene-1H-pyrimidin-4-one
Molecular Weight (g/mol) 142.18
Quantity 100 mg
Formula Weight 142.0201 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.