missing translation for 'onlineSavingsMsg'
Learn More

Methyltriphenylphosphonium Bromide, 98%

Product Code. 10031850 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
10g
2.5kg
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10031850 100 g Plastic bottle 100g
10041850 10 g Plastic bottle 10g
10319083 500 g Plastic bottle 500g
11975801 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 missing translation for 'options'
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Regulatorischer Hinweis: Abgabe sachkundepflichtiger Stoffe und Gemische nur bei Erfüllung der Vorrausetzungen nach Chemikalienverbotsverordnung (ChemVerbotsV). Keine Abgabe an Privatpersonen.
Artikelnummer. 10031850 missing translation for 'mfr' Thermo Scientific Acros missing translation for 'supplierNo' 156951000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Regulatorischer Hinweis: Abgabe sachkundepflichtiger Stoffe und Gemische nur bei Erfüllung der Vorrausetzungen nach Chemikalienverbotsverordnung (ChemVerbotsV). Keine Abgabe an Privatpersonen.

CAS: 1779-49-3 | C19H18BrP | 357.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 1779-49-3
Molecular Formula C19H18BrP
Molecular Weight (g/mol) 357.23
MDL Number MFCD00011804
InChI Key LSEFCHWGJNHZNT-UHFFFAOYSA-M
Synonym methyltriphenylphosphonium bromide, methyltriphenylphosphanium bromide, triphenylmethylphosphonium bromide, methyl triphenylphosphonium bromide, methyl triphenyl phosphonium bromide, phosphonium, methyltriphenyl-, bromide, methyltriphenylphosphoniumbromide, methyl-triphenyl-phosphonium bromide
PubChem CID 74505
IUPAC Name methyl(triphenyl)phosphanium;bromide
SMILES [Br-].C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Spezifikation

Melting Point 230.0°C to 234.0°C
Color White
Flash Point >240°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Plastic bottle
Linear Formula CH3P(C6H5)3Br
Quantity 100 g
Fieser 01,697
Solubility Information Solubility in water: 400g/L (25°C). Other solubilities: 950g/L methanol (20°C),soluble in aliphatic alcohols and chloroform,practically insoluble in arom. hydrocarbons and,acetone
Formula Weight 357.22
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Methyltriphenylphosphonium bromide
Mehr anzeigen Weniger anzeigen
compliance-icons
Produktidentifikator
  • Methyltriphenylphosphonium bromide
Signalwort
  • Warning
Gefahrenkategorie
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Sicherheitshinweis
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.