missing translation for 'onlineSavingsMsg'
Learn More

Metronidazole, Mikromol™

Product Code. 17924316
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17924316 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17924316 Supplier Mikromol Supplier No. MM0012.000250
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Metronidazole
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Metronidazole
CAS 443-48-1
Analyte or Component Names Metronidazole
Shipping Condition Room Temperature
Molecular Formula C6 H9 N3 O3
Certifications/Compliance ISO 17034
Synonym Metronidazole, Metronidazole Benzoate Imp. A (EP), 2-Methyl-5-nitro-1H-imidazole-1-ethanol, 1-(2-Hydroxyethyl)-2-methyl-5-nitroimidazole, 1-Hydroxyethyl-2-methyl-5-nitroimidazole, 2-Methyl-5-nitroimidazole-1-ethanol, Anagiardil, Arilin, Bayer 5360, Cimetrol 500LPCI, Clont, Deflamon, Efloran, Elyzol, Entizol, Flagesol, Flagil, Flagyl, Fossyol, Gineflavir, Klion, Klont, Medazol, Metro Cream & Gel, MetroGel, Metrogyl, Metrolag, Metrolyl, Metron, Metronidazol, Metropol, Metrotop, Mexibol, NSC 50364, NSC 69587, Nidazole, Noritate, Orvagil, Protostat, RP 8823, Rathimed, Sanatrichom, Satric, Takimetol, Trichazol, Tricho Cordes, Tricho-Gynaedron, Trichobrol, Trichocide, Trichomol, Trichopal, Tricocet, Trikhopol, Trivazol, Vagilen, Vagimid, Zadstat, 1-(beta-Hydroxyethyl)-2-methyl-5-nitroimidazole, Metronidazole Benzoate Impurity A
InChI Formula InChI=1S/C6H9N3O3/c1-5-7-4-6(9(11)12)8(5)2-3-10/h4,10H,2-3H2,1H3
SMILES Cc1ncc([N+](=O)[O-])n1CCO
IUPAC Name 2-(2-methyl-5-nitroimidazol-1-yl)ethanol
Molecular Weight (g/mol) 171.15
Quantity 250 mg
Formula Weight 171.0644 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.