missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METT10D Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-81239-25ul
This item is not returnable.
View return policy
Description
METT10D Polyclonal antibody specifically detects METT10D in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, KnockDown
Specifications
| METT10D | |
| Polyclonal | |
| Western Blot 0.04-0.4 ug/mL, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL, Knockdown Validated | |
| EC 2.1.1.-, methyltransferase 10 domain containing, Methyltransferase 10 domain-containing protein, methyltransferase like 16, METT10D, MGC3329, putative methyltransferase METT10D | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: DERSEEKGGVEVLESCQGSSNGAQDQEASEQFGSPVAERGKRLPGVAGQYLFKCLINVKKEVDDALVEMHWVE | |
| 25 μL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry, KnockDown | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 79066 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction