missing translation for 'onlineSavingsMsg'
Learn More
Learn More
METT11D1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
280.00 € - 589.00 €
Specifications
| Antigen | METT11D1 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18227200
|
Novus Biologicals
NBP2-54746 |
100 μL |
589.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18668506
|
Novus Biologicals
NBP2-54746-25ul |
25 μL |
280.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
METT11D1 Polyclonal specifically detects METT11D1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| METT11D1 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| EC 2.1.1.-, False p73 target gene protein, FLJ20859, methyltransferase 11 domain containing 1, Methyltransferase 11 domain-containing protein 1, methyltransferase like 17, METT11D1, Protein RSM22 homolog, mitochondrial | |
| METTL17 | |
| IgG | |
| Affinity Purified |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| 64745 | |
| This antibody was developed against a Recombinant Protein corresponding to amino acids:DPRPGFVFAPCPHELPCPQLTNLACSFSQAYHPIPFSWNKKPKEEKFSMVILARGSPEEAHRWPRITQPVLKRPRHVHCHLCCPDGHMQHAVLTARRHGRDLYRCARVSSWGDLLPVL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title