missing translation for 'onlineSavingsMsg'
Learn More

MGRN1 Antibody [FITC], Novus Biologicals Biologicals™

Product Code. 30487995 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30487995 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30487995 Supplier Novus Biologicals Supplier No. NBP337893F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MGRN1 Polyclonal antibody specifically detects MGRN1 in Mouse,Rat samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen MGRN1
Applications ELISA, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias E3 ubiquitin-protein ligase MGRN1, EC 6.3.2, KIAA0544probable E3 ubiquitin-protein ligase MGRN1, Mahogunin RING finger protein 1, mahogunin, ring finger 1, RING finger protein 156, RNF156EC 6.3.2.-
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-538 of human MGRN1 (O60291).,, Sequence:, MGSILSRRIAGVEDIDIQANSAYRYPPKSGNYFASHFFMGGEKFDTPHPEGYLFGENMDLNFLGSRPVQFPYVTPAPHEPVKTLRSLVNIRKDSLRLVRYKDDADSPTEDGDKPRVLYSLEFTFDADARVAITIYCQASEEFLNGRAVYSPKSPSLQSETVHYKRGVSQQFSLPSFKIDFSEWKDDELNFDLDRGVFPVVIQAVVDEGDVVEVTGHAHVLLAAFEKHMDGSFSVKPLKQKQIVDRVSYLLQEIYGIENKNNQETKPSDDENSDNSNECVVCLSDLRDTLILPCRHLCLCTSCADTLRYQANNCPICRLPFRALLQIRAVRKKPGALSPVSFSPVLAQSLEHDEHSCPFKKSKPHPASLASKKPKRETNSDSVPPGYEPISLLEALNGLRAVSPAIPSAPLYEEITYSGISDGLSQASCPLAAIDHILDSSRQKGRPQSKAPDSTLRSPSSPIHEEDEEKLSEDVDAPPPLGGAELALRESSSPESFITEEVDESSSPQQGTRAASIENVLQDSSPEHCGRGPPADIYL
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 23295
Target Species Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.