missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MHC Class I (H-2kb) Monoclonal Antibody (AF6-88.5.5.3), Super Bright™ 436, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
164.00 € - 470.00 €
Specifications
| Antigen | MHC Class I (H-2kb) |
|---|---|
| Clone | AF6-88.5.5.3 |
| Concentration | 0.2 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16331304
|
Affymetrix eBioscience
62-5958-80 |
25 μg | |||||||
16341304
![]() |
Invitrogen™
62-5958-82 |
100 μg |
470.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mouse Monoclonal Antibody
This protein is an MHC class I alloantigen, which is involved in antigen presentation to T cells expressing CD3/TCR and CD8. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.Specifications
| MHC Class I (H-2kb) | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| H2-K1 | |
| Primary | |
| 4° C, store in dark, DO NOT FREEZE! | |
| H2-K1 |
| AF6-88.5.5.3 | |
| Flow Cytometry | |
| Super Bright 436 | |
| Mouse | |
| Mouse | |
| 14972 | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title