Learn More
MHC Class II Antibody (ER-TR2), PerCP, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals NB100-64959PCP
488.75 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
Product Code. 18546953
  • 614.00 € / 125µL
Description
MHC Class II Monoclonal antibody specifically detects MHC Class II in Mouse samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MHC Class II
Monoclonal
PerCP
class II histocompatibility antigen, M alpha chain, D6S222E, DMA, HLA class II histocompatibility antigen, DM alpha chain, major histocompatibility complex, class II, DM alpha, MHC class II antigen DMA, Really interesting new gene 6 protein, RING6HLADM
Isolated C3H thymic stromal cells
0.125 mL
Adaptive Immunity, Asthma, B Cell Development and Differentiation Markers, Cell Biology, Diabetes Research, Immune System Diseases, Immunology, Transcription Factors and Regulators
3108
Store at 4°C in the dark.
IgG2b
Flow Cytometry
ER-TR2
PBS
Rat
Protein G purified
RUO
Primary
Mouse
Purified