missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Miconazole Nitrate, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Miconazole Nitrate |
| CAS | 22832-87-7 |
| Analyte or Component Names | Miconazole Nitrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H14 Cl4 N2 O . H N O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1-[(2RS)-2-[(2,4-Dichlorobenzyl)oxy]-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole Nitrate, Miconazole Nitrate, Isoconazole Nitrate Imp. C (EP) as Nitrate, 1-[2-(2,4-Dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]-1H-imidazole nitrate (1:1), 1-[2-(2,4-Dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]-1H-imidazole mononitrate, Aflorix, Albistat, Andergin, Antifungal Cream, Brentan, Conoderm, Conofite, Daktacort, Daktar, Daktarin, Daktarin talc, Deralbine, Dermonistat, Ecobi, Epi-Monistat, Florid, Fungiderm, Fungisdin, Gyno-Daktarin, Gyno-Monistat, Hi-Pick, Loptrimin AF Jock-Itch Powder Aerosol, Lotrimin AF Powder, Lotrimin AF Powder Aerosol, Lotrimin AF Spray Liquid, Micatin, Miconal, Miconaz, Miconazole nitrate, Micotef, Monistat, Monistat Cream and Suppositories, Monistat-Derm, NSC 169434, Priconazole, Prilagin, R 14889, Vodol, Zeasorb AF, Zimycan, 1H-Imidazole, 1-[2-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]-, mononitrate (9CI), Imidazole, 1-[2,4-dichloro-beta-[(2,4-dichlorobenzyl)oxy]phenethyl]-, mononitrate (8CI), (+/-)-Miconazole nitrate, Dactrine, [2,4-Dichloro-beta-(2,4-dichlorobenzyloxy)phenethyl]imidazole nitrate, Isoconazole Nitrate Impurity C as Nitrate, Isoconazole Impurity C as Nitrate |
| InChI Formula | InChI=1S/C18H14Cl4N2O.HNO3/c19-13-2-1-12(16(21)7-13)10-25-18(9-24-6-5-23-11-24)15-4-3-14(20)8-17(15)22;2-1(3)4/h1-8,11,18H,9-10H2;(H,2,3,4) |
| Solution Type | Neat |
| SMILES | O[N+](=O)[O-].Clc1ccc(COC(Cn2ccnc2)c3ccc(Cl)cc3Cl)c(Cl)c1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?