missing translation for 'onlineSavingsMsg'
Learn More

Migfilin Antibody (FBLIM1/8189R), Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NBP3-20310-20ug

 View more versions of this product

Product Code. 18696124

  • 286.00 € / 20µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Migfilin Monoclonal antibody specifically detects Migfilin in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Migfilin
Monoclonal
0.2 mg/ml
Immunohistochemistry-Paraffin 1-2 μg/mL
CAL, DKFZp434G171, FBLP-1FBLP1CSX-associated LIM, filamin binding LIM protein 1, filamin-binding LIM protein 1, filamin-binding LIM protein-1, MIG2-interacting protein, Migfilin, mitogen-inducible 2 interacting protein, Mitogen-inducible 2-interacting protein
Recombinant full-length human Migfilin protein
20 μg
Primary
Human
Purified
Immunohistochemistry (Paraffin)
FBLIM1/8189R
Unconjugated
10mM PBS with 0.05% BSA
Rabbit
Protein A or G purified
RUO
54751
Store at 4&drg;C. Do not freeze.
IgG κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.