missing translation for 'onlineSavingsMsg'
Learn More

Migfilin Antibody (FBLIM1/8189R), Novus Biologicals™

Rabbit Monoclonal Antibody

286.00 € - 530.00 €

Specifications

Antigen Migfilin
Clone FBLIM1/8189R
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 μg/mL
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18696124
View Documents
Novus Biologicals
NBP3-20310-20ug
20 μg
286.00 €
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18654385
View Documents
Novus Biologicals
NBP3-20310-100ug
100 μg
530.00 €
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Migfilin Monoclonal antibody specifically detects Migfilin in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

Migfilin
0.2 mg/ml
Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human
CAL, DKFZp434G171, FBLP-1FBLP1CSX-associated LIM, filamin binding LIM protein 1, filamin-binding LIM protein 1, filamin-binding LIM protein-1, MIG2-interacting protein, Migfilin, mitogen-inducible 2 interacting protein, Mitogen-inducible 2-interacting protein
Recombinant full-length human Migfilin protein
Primary
Store at 4&drg;C. Do not freeze.
FBLIM1/8189R
Immunohistochemistry-Paraffin 1-2 μg/mL
Monoclonal
Purified
RUO
10mM PBS with 0.05% BSA
54751
IgG κ
Protein A or G purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.