missing translation for 'onlineSavingsMsg'
Learn More

Milnacipran Hydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal

Product Code. 30463223
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30463223 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30463223 Supplier LoGiCal Supplier No. LGCAMP0746.0001
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Milnacipran Hydrochloride
CAS 101152-94-7
Analyte or Component Names Milnacipran Hydrochloride
Shipping Condition Ice pack (-20°C)
Molecular Formula C15 H22 N2 O . Cl H
Certifications/Compliance ISO 17034
Synonym Cyclopropanecarboxamide, 2-(aminomethyl)-N,N-diethyl-1-phenyl-, hydrochloride (1:1), Cyclopropanecarboxamide, 2-(aminomethyl)-N,N-diethyl-1-phenyl-, monohydrochloride, (1R,2S)-rel- (9CI), Cyclopropanecarboxamide, 2-(aminomethyl)-N,N-diethyl-1-phenyl-, monohydrochloride, cis-, Dalcipran, F 2207, Ixel, Joncia, Milnacipran hydrochloride, Savella, Tivanyl, Toledomin, (1R,2S)-rel-, rac-Milnacipran Hydrochloride, Cyclopropanecarboxamide, 2-(aminomethyl)-N,N-diethyl-1-phenyl-, monohydrochloride, cis-(+/-)-
InChI Formula InChI=1S/C15H22N2O.ClH/c1-3-17(4-2)14(18)15(10-13(15)11-16)12-8-6-5-7-9-12;/h5-9,13H,3-4,10-11,16H2,1-2H3;1H/t13-,15+;/m0./s1
Solution Type Single Solution
SMILES Cl.CCN(CC)C(=O)[C@]1(C[C@H]1CN)c2ccccc2
IUPAC Name (1S,2R)-2-(aminomethyl)-N,N-diethyl-1-phenylcyclopropane-1-carboxamide;hydrochloride
Solvent or Matrix Methanol
Industry Type Food Testing
Molecular Weight (g/mol) 282.81
Quantity 1 mL
Formula Weight 282.1499 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.