missing translation for 'onlineSavingsMsg'
Learn More

MMS19 like protein Antibody (6H1) [mFluor Violet 610 SE], Novus Biologicals Biologicals™

Product Code. 30520987 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30520987 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30520987 Supplier Novus Biologicals Supplier No. NBP343000MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MMS19 like protein Monoclonal antibody specifically detects MMS19 like protein in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen MMS19 like protein
Applications ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone 6H1
Conjugate mFluor Violet 610 SE
Formulation 50mM Sodium Borate
Gene Alias FLJ34167, MET18, MET18 homolog, MMS19 nucleotide excision repair homolog (S. cerevisiae), S. cerevisiae)
Host Species Mouse
Immunogen MMS19 nucleotide excision repair homolog(S. cerevisiae) - (Uniprot# Q96T76)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Direct Reversal of DNA Damage, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 64210
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.