missing translation for 'onlineSavingsMsg'
Learn More

MOK Antibody - BSA Free, Novus Biologicals™

Product Code. 18672722 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.02 mL
0.1 mL
Unit Size:
0.01mL
0.02mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18672722 0.02 mL 0.02mL
18631272 0.1 mL 0.01mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18672722 Supplier Novus Biologicals Supplier No. NBP2944480.02ml

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MOK Polyclonal antibody specifically detects MOK in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen MOK
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin
Formulation PBS (pH 7.3), 50% glycerol
Gene Alias EC 2.7.11.22, MAPK/MAK/MRK overlapping kinase, MOK Protein Kinase, RAGE, RAGE1, RAGE-1, renal cell carcinoma antigen, Renal tumor antigen 1, STK30
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-230 of human MOK (NP_055041.1). MKNYKAIGKIGEGTFSEVMKMQSLRDGNYYACKQMKQRFESIEQVNNLREIQALRRLNPHPNILMLHEVVFDRKSGSLALICELMDMNIYELIRGRRYPLSEKKIMHYMYQLCKSLDHIHRNGIFHRDVKPENILIKQDVLKLGDFGSCRSVYSKQPYTEYISTRWYRAPECLLTDGFYTYKMDLWSAGCVFYEIASLQPLFPGVNELDQISKIHDVIGTPAQKILTKFK
Purification Method Affinity purified
Quantity 0.02 mL
Regulatory Status RUO
Research Discipline Neurodegeneration, Neuronal Cell Markers, Neuroscience, Protein Kinase, Vision
Primary or Secondary Primary
Gene ID (Entrez) 5891
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.