missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monkeypox Virus A35 Antibody (0026), mFluor Violet 450 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Monkeypox Virus A35 Monoclonal antibody specifically detects Monkeypox Virus A35 in Virus samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | Monkeypox Virus A35 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 0026 |
| Conjugate | mFluor Violet 450 SE |
| Dilution | Western Blot, ELISA |
| Formulation | 50mM Sodium Borate |
| Host Species | Mouse |
| Immunogen | This product is a recombinant monoclonal antibody made to recombinant Monkeypox Virus A35 and expressed from HEK293 cells. |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?