Learn More
Monkeypox Virus A35 Antibody (0026), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-18812MFV610
555.90 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
Product Code. 18528527
  • 650.00 € / 0.10mL
Description
Monkeypox Virus A35 Monoclonal antibody specifically detects Monkeypox Virus A35 in Virus samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Monkeypox Virus A35
Monoclonal
mFluor Violet 610 SE
50mM Sodium Borate
This product is a recombinant monoclonal antibody made to recombinant Monkeypox Virus A35 and expressed from HEK293 cells.
0.1 mL
Primary
Store at 4C in the dark.
IgG1
Western Blot, ELISA
0026
Western Blot, ELISA
Mouse
Protein A purified
RUO
Virus
Purified