Learn More
Invitrogen™ Monkeypox Virus B6R Recombinant Rabbit Monoclonal Antibody (006), HRP
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA560392
 View more versions of this product
Product Code. 30534273
  • 209.00 € / 20µg
Description
Monkeypox Virus B6R Recombinant Monoclonal Antibody for ELISA
The monkeypox virus (MPXV) is the causative agent of the monkeypox disease. As a double-stranded DNA virus member of the genus Orthopoxvirus and the family Poxviridae, MPXV is similar to other pathogens of this group including variola (the causative agent of smallpox), vaccinia, and the cowpox virus. MPXV A29L, M1R and B6R, the orthologous to A27L, L1R and B5R VACV antigens respectively, are thought to be able to elicit a protective immune response against lethal MPXV challenge.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Monkeypox Virus B6R
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
Rabbit
Protein A
RUO
Virus
Antibody
IgG
ELISA
006
HRP
Monkeypox B6R
Recombinant Monkeypox Virus (MPXV) B6R protein (Tyr18-His279).
20 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles