missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Monkeypox Virus B6R Recombinant Rabbit Monoclonal Antibody (006), HRP

Rabbit Recombinant Monoclonal Antibody
209.00 € - 569.00 €
Specifications
| Antigen | Monkeypox Virus B6R |
|---|---|
| Clone | 006 |
| Concentration | 1 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30534607
![]() |
Invitrogen™
MA560391 |
100 μg |
569.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30534273
|
Invitrogen™
MA560392 |
20 μg |
209.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Monkeypox Virus B6R Recombinant Monoclonal Antibody for ELISA
The monkeypox virus (MPXV) is the causative agent of the monkeypox disease. As a double-stranded DNA virus member of the genus Orthopoxvirus and the family Poxviridae, MPXV is similar to other pathogens of this group including variola (the causative agent of smallpox), vaccinia, and the cowpox virus. MPXV A29L, M1R and B6R, the orthologous to A27L, L1R and B5R VACV antigens respectively, are thought to be able to elicit a protective immune response against lethal MPXV challenge.Specifications
| Monkeypox Virus B6R | |
| 1 mg/mL | |
| ELISA | |
| HRP | |
| Rabbit | |
| RUO | |
| PBS with no preservative | |
| Recombinant Monkeypox Virus (MPXV) B6R protein (Tyr18-His279). | |
| Antibody |
| 006 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Virus | |
| Monkeypox B6R | |
| Primary | |
| Protein A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title