missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Monoethyl Fumarate Calcium, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Monoethyl Fumarate Calcium |
| CAS | 62008-22-4 |
| Analyte or Component Names | Monoethyl Fumarate Calcium |
| Shipping Condition | Room Temperature |
| Molecular Formula | 2 C6 H7 O4 . Ca |
| Certifications/Compliance | ISO 9001 |
| Synonym | Monoethyl Fumarate Calcium, 2-Butenedioic acid (2E)-, 1-ethyl ester, calcium salt (2:1), 2-Butenedioic acid (2E)-, monoethyl ester, calcium salt (9CI), 2-Butenedioic acid (E)-, monoethyl ester, calcium salt, Calcium monoethylfumarate, Monoethyl fumarate calcium salt |
| InChI Formula | InChI=1S/2C6H8O4.Ca/c2*1-2-10-6(9)4-3-5(7)8;/h2*3-4H,2H2,1H3,(H,7,8);/q;;+2/p-2/b2*4-3+; |
| Solution Type | Neat |
| SMILES | [Ca+2].CCOC(=O)\C=C\C(=O)[O-].CCOC(=O)\C=C\C(=O)[O-] |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?