missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Aminopeptidase N/CD13 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB2335
This item is not returnable.
View return policy
Description
Aminopeptidase N/CD13 Monoclonal specifically detects Aminopeptidase N/CD13 in Mouse samples. It is validated for Western Blot, Simple Western.
Specifications
| Aminopeptidase N/CD13 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| alanyl (membrane) aminopeptidase, Alanyl aminopeptidase, Aminopeptidase M, aminopeptidase N, ANPEP, AP-M, APN, AP-N, CD13, CD13 antigen, CD13APN, EC 3.4.11, EC 3.4.11.2, gp150, LAP1, Microsomal aminopeptidase, Myeloid plasma membrane glycoprotein CD13, P150, PEPN, PEPNhAPN | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 290 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot | |
| 945033 | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL | |
| P97449 | |
| Anpep | |
| NS0 mouse myeloma cell line transfected with mouse Aminopeptidase Lys69-Ser966 Accession # P97449 | |
| 100 μg | |
| Primary | |
| Detects mouse Aminopeptidase in direct ELISAs and Western blots. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction