missing translation for 'onlineSavingsMsg'
Learn More

Mouse Aminopeptidase N/CD13 Antibody, R&D Systems™
GREENER_CHOICE

Rat Monoclonal Antibody

184.00 € - 377.00 €

Specifications

Antigen Aminopeptidase N/CD13
Clone 945033
Dilution Western Blot 1 ug/mL, Simple Western 10 ug/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18779668
GREENER_CHOICE
View Documents
R&D Systems
MAB2335-SP
25 μg
184.00 €
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18755779
GREENER_CHOICE
View Documents
R&D Systems
MAB2335
100 μg
377.00 €
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Aminopeptidase N/CD13 Monoclonal specifically detects Aminopeptidase N/CD13 in Mouse samples. It is validated for Western Blot, Simple Western.
Specifications

Specifications

Aminopeptidase N/CD13
Western Blot 1 ug/mL, Simple Western 10 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
alanyl (membrane) aminopeptidase, Alanyl aminopeptidase, Aminopeptidase M, aminopeptidase N, ANPEP, AP-M, APN, AP-N, CD13, CD13 antigen, CD13APN, EC 3.4.11, EC 3.4.11.2, gp150, LAP1, Microsomal aminopeptidase, Myeloid plasma membrane glycoprotein CD13, P150, PEPN, PEPNhAPN
Anpep
IgG2a
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
945033
Western Blot
Unconjugated
Rat
Mouse
P97449
290
NS0 mouse myeloma cell line transfected with mouse Aminopeptidase Lys69-Ser966 Accession # P97449
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects mouse Aminopeptidase in direct ELISAs and Western blots.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.