missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human BDNF Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
214.00 € - 2090.00 €
Specifications
| Antigen | BDNF |
|---|---|
| Clone | 37141 |
| Dilution | Western Blot 1 ug/mL, ELISA |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18776249
![]() |
R&D Systems
MAB648-SP |
25 μg |
214.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18747778
![]() |
R&D Systems
MAB648-100 |
100 μg |
517.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18768478
![]() |
R&D Systems
MAB648-500 |
500 μg |
2090.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BDNF Monoclonal specifically detects BDNF in Human samples. It is validated for Western Blot, ELISA.Specifications
| BDNF | |
| Western Blot 1 ug/mL, ELISA | |
| Monoclonal | |
| Mouse | |
| Human | |
| P23560 | |
| 627 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human BDNF Arg128-Arg247 Accession # P23560 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human BDNF in direct ELISAs and Western blots. Does not cross-react with recombinant human (rh) NT-3 or rhNT-4. |
| 37141 | |
| Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| abrineurin, ANON2, brain-derived neurotrophic factor, BULN2, MGC34632, neurotrophin | |
| BDNF | |
| IgG2a | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title