Learn More
Mouse CCL6/C10 Antibody, R&D Systems™
GREENER_CHOICE
Rat Monoclonal Antibody
Brand:  R&D Systems MAB4872-100
 View more versions of this product
Product Code. 18743347
  • 440.00 € / 100µg
Description
CCL6/C10 Monoclonal specifically detects CCL6/C10 in Mouse samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CCL6/C10
Monoclonal
Unconjugated
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
20305
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
IgG2a
ELISA
262008
ELISA
P27784
E. coli-derived recombinant mouse CCL6/C10, Gly22-Ala116, Accession # P27784
100 μg
Primary
Detects mouse CCL6/C10 in direct ELISAs.
Mouse
Purified