missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CCL6/C10 Antibody, R&D Systems™

Rat Monoclonal Antibody
176.00 € - 440.00 €
Specifications
| Antigen | CCL6/C10 |
|---|---|
| Clone | 262008 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18773997
![]() |
R&D Systems
MAB4872-SP |
25 μg |
176.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18743347
![]() |
R&D Systems
MAB4872-100 |
100 μg |
440.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCL6/C10 Monoclonal specifically detects CCL6/C10 in Mouse samples. It is validated for ELISA.Specifications
| CCL6/C10 | |
| ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| 20305 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| 262008 | |
| ELISA | |
| Unconjugated | |
| Rat | |
| Mouse | |
| P27784 | |
| E. coli-derived recombinant mouse CCL6/C10, Gly22-Ala116, Accession # P27784 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse CCL6/C10 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title