missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CCL6/C10 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB4872-SP
This item is not returnable.
View return policy
Description
CCL6/C10 Monoclonal specifically detects CCL6/C10 in Mouse samples. It is validated for ELISA.
Specifications
| CCL6/C10 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 20305 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2a |
| ELISA | |
| 262008 | |
| ELISA | |
| P27784 | |
| E. coli-derived recombinant mouse CCL6/C10, Gly22-Ala116, Accession # P27784 | |
| 25 μg | |
| Primary | |
| Detects mouse CCL6/C10 in direct ELISAs. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction