missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CCR9 PE-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems FAB2160P-100
This item is not returnable.
View return policy
Description
CCR9 Monoclonal specifically detects CCR9 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| CCR9 | |
| Monoclonal | |
| R-PE | |
| Q9WUT7 | |
| CCR9 | |
| Y3 rat myeloid cell line transfected with mouse CCR9 Met1-Leu369 Accession # Q9WUT7 | |
| 100 Tests | |
| 10803 | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 242503 | |
| Flow Cytometry 10 uL/10^6 cells | |
| C-C chemokine receptor type 9, C-C CKR-9, CC-CKR-9, CCR9, CCR-9, CD199, CDw199, CDw199 antigen, chemokine (C-C motif) receptor 9, G protein-coupled receptor 28, GPR28, GPR-9-6G-protein coupled receptor 28 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects mouse CCR9 in flow cytometry. Stains mouse CCR9-transfected cells but not irrelevant transfectants. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction