missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CCR9 PE-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 3 publications
200.00 € - 492.00 €
Specifications
| Antigen | CCR9 |
|---|---|
| Clone | 242503 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18771018
![]() |
R&D Systems
FAB2160P-025 |
25 Tests |
200.00 €
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18728216
![]() |
R&D Systems
FAB2160P-100 |
100 Tests |
492.00 €
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCR9 Monoclonal specifically detects CCR9 in Mouse samples. It is validated for Flow Cytometry.Specifications
| CCR9 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| Mouse | |
| C-C chemokine receptor type 9, C-C CKR-9, CC-CKR-9, CCR9, CCR-9, CD199, CDw199, CDw199 antigen, chemokine (C-C motif) receptor 9, G protein-coupled receptor 28, GPR28, GPR-9-6G-protein coupled receptor 28 | |
| CCR9 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects mouse CCR9 in flow cytometry. Stains mouse CCR9-transfected cells but not irrelevant transfectants. |
| 242503 | |
| Flow Cytometry | |
| R-PE | |
| Rat | |
| Q9WUT7 | |
| 10803 | |
| Y3 rat myeloid cell line transfected with mouse CCR9 Met1-Leu369 Accession # Q9WUT7 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title