missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD45 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 8 publications
Brand: R&D Systems MAB114-100
This item is not returnable.
View return policy
Description
CD45 Monoclonal specifically detects CD45 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Complement-dependent Cytotoxicity, Immunocytochemistry, CyTOF-reported.
Specifications
| CD45 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| PTPRC | |
| Mouse thymus and spleen cells | |
| 100 μg | |
| Primary | |
| Detects mouse CD45. Recognizes all CD45 isoforms as well as the known mouse CD45 alloantigens (CD45-1/Ly 5a and CD45-2/Ly 5b) (15). | |
| Mouse | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Cytotoxicity Assay, Immunocytochemistry | |
| 30-F11 | |
| Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-25 ug/mL, Immunoprecipitation, Complement-dependent Cytotoxicity, Immunocytochemistry 3-25 ug/mL, CyTOF-reported | |
| B220, CD45, CD45 antigen, CD45R, EC 3.1.3.48, LCA, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, PTPRC, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5788 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction