missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse CD45 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 8 publications
224.00 € - 2040.00 €
Specifications
| Antigen | CD45 |
|---|---|
| Clone | 30-F11 |
| Dilution | Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-25 ug/mL, Immunoprecipitation, Complement-dependent Cytotoxicity, Immunocytochemistry 3-25 ug/mL, CyTOF-reported |
| Applications | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Cytotoxicity Assay, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18711647
![]() |
R&D Systems
MAB114-SP |
25 μg |
224.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18739517
![]() |
R&D Systems
MAB114-100 |
100 μg |
512.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18707569
![]() |
R&D Systems
MAB114-500 |
500 μg |
2040.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD45 Monoclonal specifically detects CD45 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Complement-dependent Cytotoxicity, Immunocytochemistry, CyTOF-reported.Specifications
| CD45 | |
| Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 5-25 ug/mL, Immunoprecipitation, Complement-dependent Cytotoxicity, Immunocytochemistry 3-25 ug/mL, CyTOF-reported | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 5788 | |
| Mouse thymus and spleen cells | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse CD45. Recognizes all CD45 isoforms as well as the known mouse CD45 alloantigens (CD45-1/Ly 5a and CD45-2/Ly 5b) (15). |
| 30-F11 | |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Cytotoxicity Assay, Immunocytochemistry | |
| Unconjugated | |
| Rat | |
| Mouse | |
| B220, CD45, CD45 antigen, CD45R, EC 3.1.3.48, LCA, L-CA, LY5, protein tyrosine phosphatase, receptor type, C, PTPRC, receptor-type tyrosine-protein phosphatase C, T200 glycoprotein, T200 leukocyte common antigen, T200receptor type, c polypeptide | |
| PTPRC | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title