missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Cripto Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB15381-SP
This item is not returnable.
View return policy
Description
CRIPTO Monoclonal specifically detects CRIPTO in Mouse samples. It is validated for Western Blot.
Specifications
| CRIPTO | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CFC-2, CR, Cripto-1, Cripto-1 growth factor, CRIPTOCRGF, Epidermal growth factor-like cripto protein CR1, TDGF1, TDGF-1, teratocarcinoma-derived growth factor 1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6997 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot | |
| 237614 | |
| Western Blot 1 ug/mL | |
| P51865 | |
| Tdgf1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Cripto Arg26-Gln150 Accession # P51865 | |
| 25 μg | |
| Primary | |
| Detects mouse Cripto in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) ART, rmArtemin, recombinant human (rh) Cripto-1, recombinant drosophilia DPP, recombinant rat (rr) GDNF, rhLAP, rmLefty-1, rrMIS, rhNeurturin, rmNodal, rhTGF-alpha , rhTGF-beta 1, rhTGF-beta 1.2, rhTGF-beta 2, rhTGF-beta 3, or recombinant amphibian TGF-beta 5 is observed. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction