missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Cripto Antibody, R&D Systems™

Rat Monoclonal Antibody
188.00 € - 496.00 €
Specifications
| Antigen | CRIPTO |
|---|---|
| Clone | 237614 |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18762937
![]() |
R&D Systems
MAB15381-SP |
25 μg |
188.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18139350
![]() |
R&D Systems
MAB15381 |
500 μg |
496.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CRIPTO Monoclonal specifically detects CRIPTO in Mouse samples. It is validated for Western Blot.Specifications
| CRIPTO | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CFC-2, CR, Cripto-1, Cripto-1 growth factor, CRIPTOCRGF, Epidermal growth factor-like cripto protein CR1, TDGF1, TDGF-1, teratocarcinoma-derived growth factor 1 | |
| Tdgf1 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 237614 | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Mouse | |
| P51865 | |
| 6997 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Cripto Arg26-Gln150 Accession # P51865 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse Cripto in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) ART, rmArtemin, recombinant human (rh) Cripto-1, recombinant drosophilia DPP, recombinant rat (rr) GDNF, rhLAP, rmLefty-1, rrMIS, rhNeurturin, rmNodal, rhTGF-alpha , rhTGF-beta 1, rhTGF-beta 1.2, rhTGF-beta 2, rhTGF-beta 3, or recombinant amphibian TGF-beta 5 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title