missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse GDF-5/BMP-14 Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB853
This item is not returnable.
View return policy
Description
GDF-5/BMP-14 Monoclonal specifically detects GDF-5/BMP-14 in Mouse samples. It is validated for Western Blot.
Specifications
| GDF-5/BMP-14 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BMP14, BMP-14, Cartilage-derived morphogenetic protein 1, cartilage-derived morphogenetic protein-1, CDMP-1, CDMP1LAP4, GDF5, GDF-5, growth differentiation factor 5, growth/differentiation factor 5, OS5, Radotermin, SYNS2 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8200 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot | |
| 90509 | |
| Western Blot 1 ug/mL | |
| P43027 | |
| GDF5 | |
| E. coli-derived recombinant mouse GDF-5/BMP-14 Ala376-Arg495 Accession # P43027 | |
| 500 μg | |
| Primary | |
| Detects mouse GDF-5/BMP-14 in direct ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with recombinant mouse (rm) GDF-7 and approximately 50% cross-reactivity with rmGDF-6 is observed. Less than 10% cross-reactivity with rmGDF-3 (dimer), -6, -7, -8, -9, -11, -15 (dimer), rmPro-GDF-1, rmPro-GDF-3, or rmPro-GDF-9 is observed. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction