missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse GDF-5/BMP-14 Antibody, R&D Systems™

Rat Monoclonal Antibody
196.00 € - 500.00 €
Specifications
| Antigen | GDF-5/BMP-14 |
|---|---|
| Clone | 90509 |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18738879
![]() |
R&D Systems
MAB853-SP |
25 μg |
196.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18129290
|
R&D Systems
MAB853 |
500 μg |
500.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GDF-5/BMP-14 Monoclonal specifically detects GDF-5/BMP-14 in Mouse samples. It is validated for Western Blot.Specifications
| GDF-5/BMP-14 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BMP14, BMP-14, Cartilage-derived morphogenetic protein 1, cartilage-derived morphogenetic protein-1, CDMP-1, CDMP1LAP4, GDF5, GDF-5, growth differentiation factor 5, growth/differentiation factor 5, OS5, Radotermin, SYNS2 | |
| GDF5 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 90509 | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Mouse | |
| P43027 | |
| 8200 | |
| E. coli-derived recombinant mouse GDF-5/BMP-14 Ala376-Arg495 Accession # P43027 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse GDF-5/BMP-14 in direct ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with recombinant mouse (rm) GDF-7 and approximately 50% cross-reactivity with rmGDF-6 is observed. Less than 10% cross-reactivity with rmGDF-3 (dimer), -6, -7, -8, -9, -11, -15 (dimer), rmPro-GDF-1, rmPro-GDF-3, or rmPro-GDF-9 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title