missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IFN-gamma APC-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 1 publication
Brand: R&D Systems IC485A-100
This item is not returnable.
View return policy
Description
IFN-gamma Monoclonal specifically detects IFN-gamma in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| IFN-gamma | |
| Monoclonal | |
| APC | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| IFNG | |
| E. coli-derived recombinant mouse IFN-gamma | |
| 100 Tests | |
| Primary | |
| Detects mouse IFN-gamma in Western blots. In Western blots, this antibody does not cross-react with recombinant human (rh) IFN-gamma , rrIFN-gamma , rpIFN-gamma , rrmIFN-gamma , rfeIFN-gamma , or rcaIFN-gamma . | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2a |
| Flow Cytometry | |
| 37895 | |
| Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
| IFG, IFI, IFNG, IFNgamma, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3458 | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction