missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IFN-gamma APC-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 1 publication
262.00 € - 514.00 €
Specifications
| Antigen | IFN-gamma |
|---|---|
| Clone | 37895 |
| Dilution | Intracellular Staining by Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18788398
![]() |
R&D Systems
IC485A-025 |
25 Tests |
262.00 €
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18721927
![]() |
R&D Systems
IC485A-100 |
100 Tests |
514.00 €
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IFN-gamma Monoclonal specifically detects IFN-gamma in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
| IFN-gamma | |
| Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| 3458 | |
| E. coli-derived recombinant mouse IFN-gamma | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| 37895 | |
| Flow Cytometry | |
| APC | |
| Rat | |
| Mouse | |
| IFG, IFI, IFNG, IFNgamma, IFN-gamma, Immune interferon, interferon gamma, interferon, gamma | |
| IFNG | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects mouse IFN-gamma in Western blots. In Western blots, this antibody does not cross-react with recombinant human (rh) IFN-gamma , rrIFN-gamma , rpIFN-gamma , rrmIFN-gamma , rfeIFN-gamma , or rcaIFN-gamma . |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title