missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-21 APC-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 1 publication
Brand: R&D Systems IC594A-025
This item is not returnable.
View return policy
Description
IL-21 Monoclonal specifically detects IL-21 in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| IL-21 | |
| Monoclonal | |
| APC | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CVID11, IL21, IL-21Za11interleukin-21, interleukin 21, interleukin-21 isoform, Za11 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 59067 | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 149204 | |
| Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
| Q9ES17 | |
| IL21 | |
| E. coli-derived recombinant mouse IL-21 Pro25-Ser146 Accession # Q9ES17 | |
| 25 Tests | |
| Primary | |
| Detects mouse IL-21 in Western blots. Does not cross-react with recombinant human IL-21, recombinant mouse (rm) IL-15, or rmIL-12. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction