missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-21 APC-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 1 publication
304.00 € - 536.00 €
Specifications
| Antigen | IL-21 |
|---|---|
| Clone | 149204 |
| Dilution | Intracellular Staining by Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18774319
![]() |
R&D Systems
IC594A-025 |
25 Tests |
304.00 €
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18761717
![]() |
R&D Systems
IC594A-100 |
100 Tests |
536.00 €
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-21 Monoclonal specifically detects IL-21 in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
| IL-21 | |
| Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CVID11, IL21, IL-21Za11interleukin-21, interleukin 21, interleukin-21 isoform, Za11 | |
| IL21 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects mouse IL-21 in Western blots. Does not cross-react with recombinant human IL-21, recombinant mouse (rm) IL-15, or rmIL-12. |
| 149204 | |
| Flow Cytometry | |
| APC | |
| Rat | |
| Mouse | |
| Q9ES17 | |
| 59067 | |
| E. coli-derived recombinant mouse IL-21 Pro25-Ser146 Accession # Q9ES17 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title