missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-23R PE-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems FAB16861P-100UG
This item is not returnable.
View return policy
Description
IL-23R Monoclonal specifically detects IL-23R in Mouse samples. It is validated for Flow Cytometry.
Specifications
| IL-23R | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| IBD17, IL-23 R, IL-23 receptor, IL23R, IL-23R, interleukin 23 receptor, interleukin-23 receptor | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects endogenous mouse IL-23 R by flow cytometry. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG1 |
| Flow Cytometry | |
| 753317 | |
| Flow Cytometry 0.25-1 μg/106 cells | |
| Q8K4B4 | |
| IL23R | |
| Mouse myeloma cell line NS0-derived recombinant mouse IL-23 R, Gly24-Asp372, Accession No. Q8K4B4 | |
| 100 μg | |
| 149233.0 | |
| Mouse | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction