missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse IL-23R PE-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
268.00 € - 456.00 €
Specifications
| Antigen | IL-23R |
|---|---|
| Clone | 753317 |
| Dilution | Flow Cytometry 0.25-1 μg/106 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18759758
![]() |
R&D Systems
FAB16861P-025UG |
25 μg |
268.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18702458
![]() |
R&D Systems
FAB16861P-100UG |
100 μg |
456.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-23R Monoclonal specifically detects IL-23R in Mouse samples. It is validated for Flow Cytometry.Specifications
| IL-23R | |
| Flow Cytometry 0.25-1 μg/106 cells | |
| Monoclonal | |
| Supernatant | |
| Mouse | |
| Q8K4B4 | |
| 149233.0 | |
| Mouse myeloma cell line NS0-derived recombinant mouse IL-23 R, Gly24-Asp372, Accession No. Q8K4B4 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. |
| 753317 | |
| Flow Cytometry | |
| R-PE | |
| Rat | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| IBD17, IL-23 R, IL-23 receptor, IL23R, IL-23R, interleukin 23 receptor, interleukin-23 receptor | |
| IL23R | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects endogenous mouse IL-23 R by flow cytometry. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title