Learn More
Mouse TLR3 Antibody, R&D Systems™
GREENER_CHOICE
Rat Monoclonal Antibody has been used in 2 publications
Brand:  R&D Systems MAB3005
 View more versions of this product
Product Code. 18111661
  • 427.00 € / 100µg
Description
TLR3 Monoclonal specifically detects TLR3 in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TLR3
Monoclonal
LYOPH
Western Blot 1 ug/mL
Q99MB1
TLR3
Mouse myeloma cell line NS0-derived recombinant mouse TLR3 Cys29-Leu705 Accession # Q99MB1
100 μg
Primary
Detects mouse TLR3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse TLR1, 2, 4, 6, recombinant human TLR3 or 4 is observed.
Mouse
Purified
Western Blot
313129
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
CD283, CD283 antigen, toll-like receptor 3
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
7098
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2a