missing translation for 'onlineSavingsMsg'
Learn More

Mouse TLR3 Antibody, R&D Systems™
GREENER_CHOICE

Rat Monoclonal Antibody has been used in 2 publications

208.00 € - 427.00 €

Specifications

Antigen TLR3
Clone 313129
Concentration LYOPH
Dilution Western Blot 1 ug/mL
Applications Western Blot
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18719038
GREENER_CHOICE
View Documents
R&D Systems
MAB3005-SP
25 μg
208.00 €
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18111661
GREENER_CHOICE
View Documents
R&D Systems
MAB3005
100 μg
427.00 €
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TLR3 Monoclonal specifically detects TLR3 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

TLR3
LYOPH
Western Blot
Unconjugated
Rat
Mouse
Q99MB1
7098
Mouse myeloma cell line NS0-derived recombinant mouse TLR3 Cys29-Leu705 Accession # Q99MB1
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects mouse TLR3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse TLR1, 2, 4, 6, recombinant human TLR3 or 4 is observed.
313129
Western Blot 1 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
CD283, CD283 antigen, toll-like receptor 3
TLR3
IgG2a
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.