missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TWEAK/TNFSF12 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB1237-SP
This item is not returnable.
View return policy
Description
TWEAK/TNFSF12 Monoclonal specifically detects TWEAK/TNFSF12 in Mouse samples. It is validated for ELISA Capture (Matched Antibody Pair).
Specifications
| TWEAK/TNFSF12 | |
| Monoclonal | |
| LYOPH | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| O54907.2 | |
| Tnfsf12 | |
| E. coli-derived recombinant mouse TWEAK/TNFSF12 Arg105-His249 Accession # O54907.2 | |
| 25 μg | |
| Primary | |
| Detects mouse TWEAK/TNFSF12 in ELISAs. In sandwich ELISAs, approximately 5% cross-reactivity with recombinant human TWEAK is observed and no cross-reactivity with recombinant mouse (rm) BAFF, rmFas Ligand, rmOX40, or rmTNF-alpha is observed. | |
| Mouse | |
| Purified |
| ELISA | |
| 189803 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| APO3 ligand, APO3LMGC20669, DR3LGTWEAKAPO3/DR3 ligand, MGC129581, TNF-related weak inducer of apoptosis, TNFSF12, tumor necrosis factor (ligand) superfamily, member 12, tumor necrosis factor ligand superfamily member 12 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8742 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction