missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse TWEAK/TNFSF12 Antibody, R&D Systems™

Rat Monoclonal Antibody
188.00 € - 506.00 €
Specifications
| Antigen | TWEAK/TNFSF12 |
|---|---|
| Clone | 189803 |
| Concentration | LYOPH |
| Dilution | ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18753919
![]() |
R&D Systems
MAB1237-SP |
25 μg |
188.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18146021
![]() |
R&D Systems
MAB1237 |
500 μg |
506.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TWEAK/TNFSF12 Monoclonal specifically detects TWEAK/TNFSF12 in Mouse samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
| TWEAK/TNFSF12 | |
| LYOPH | |
| ELISA | |
| Unconjugated | |
| Rat | |
| Mouse | |
| O54907.2 | |
| 8742 | |
| E. coli-derived recombinant mouse TWEAK/TNFSF12 Arg105-His249 Accession # O54907.2 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse TWEAK/TNFSF12 in ELISAs. In sandwich ELISAs, approximately 5% cross-reactivity with recombinant human TWEAK is observed and no cross-reactivity with recombinant mouse (rm) BAFF, rmFas Ligand, rmOX40, or rmTNF-alpha is observed. |
| 189803 | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| APO3 ligand, APO3LMGC20669, DR3LGTWEAKAPO3/DR3 ligand, MGC129581, TNF-related weak inducer of apoptosis, TNFSF12, tumor necrosis factor (ligand) superfamily, member 12, tumor necrosis factor ligand superfamily member 12 | |
| Tnfsf12 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title