missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mre11 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-95102-0.1ml
This item is not returnable.
View return policy
Description
Mre11 Polyclonal antibody specifically detects Mre11 in Human, Rat samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| Mre11 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, Immunoprecipitation 1:500 - 1:1000 | |
| ATLDmeiotic recombination (S. cerevisiae) 11 homolog A, AT-like disease, DNA recombination and repair protein, double-strand break repair protein MRE11A, endo/exonuclease Mre11, HNGS1, Meiotic recombination 11 homolog 1, Meiotic recombination 11 homolog A, MRE11 homolog 1, MRE11 homolog A, MRE11 meiotic recombination 11 homolog A (S. cerevisiae), MRE11B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 290-485 of human MRE11 (NP_005582.1). KGRKMNMHKIPLHTVRQFFMEDIVLANHPDIFNPDNPKVTQAIQSFCLEKIEEMLENAERERLGNSHQPEKPLVRLRVDYSGGFEPFSVLRFSQKFVDRVANPKDIIHFFRHREQKEKTGEEINFGKLITKPSEGTTLRVEDLVKQYFQTAEKNVQLSLLTERGMGEAVQEFVDKEEKDAIEELVKYQLEKTQRFL | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Non homologous end joining, Protein Phosphatase | |
| 4361 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction