missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mre11 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
188.00 € - 470.00 €
Specifications
| Antigen | Mre11 |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, Immunoprecipitation 1:500 - 1:1000 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18640902
|
Novus Biologicals
NBP2-95102-0.02ml |
0.02 mL |
188.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18666102
|
Novus Biologicals
NBP2-95102-0.1ml |
0.1 mL |
470.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mre11 Polyclonal antibody specifically detects Mre11 in Human, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| Mre11 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Non homologous end joining, Protein Phosphatase | |
| PBS (pH 7.3), 50% glycerol | |
| 4361 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:1000, Immunoprecipitation 1:500 - 1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| ATLDmeiotic recombination (S. cerevisiae) 11 homolog A, AT-like disease, DNA recombination and repair protein, double-strand break repair protein MRE11A, endo/exonuclease Mre11, HNGS1, Meiotic recombination 11 homolog 1, Meiotic recombination 11 homolog A, MRE11 homolog 1, MRE11 homolog A, MRE11 meiotic recombination 11 homolog A (S. cerevisiae), MRE11B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 290-485 of human MRE11 (NP_005582.1). KGRKMNMHKIPLHTVRQFFMEDIVLANHPDIFNPDNPKVTQAIQSFCLEKIEEMLENAERERLGNSHQPEKPLVRLRVDYSGGFEPFSVLRFSQKFVDRVANPKDIIHFFRHREQKEKTGEEINFGKLITKPSEGTTLRVEDLVKQYFQTAEKNVQLSLLTERGMGEAVQEFVDKEEKDAIEELVKYQLEKTQRFL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title