missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MRPL48 Polyclonal antibody specifically detects MRPL48 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | MRPL48 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000 |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | 39S ribosomal protein L48, mitochondrial, CGI-118, FLJ17047, FLJ99260, HSPC290, L48MT, MGC13323, mitochondrial ribosomal protein L48, MRP-L48 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-212 of human MRPL48 (NP_057139.1). MSGTLEKVLCLRNNTIFKQAFSLLRFRTSGEKPIYSVGGILLSISRPYKTKPTHGIGKYKHLIKAEEPKKKKGKVEVRAINLGTDYEYGVLNIHLTAYDMTLAESYAQYVHNLCNSLSIKVEESYAMPTKTIEVLQLQDQGSKMLLDSVLTTHERVVQISGLSATFAEIFLEIIQSSLPEGVRLSVKEHTEEDFKGRFKARPELEELLAKLK |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?