missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS28 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00 € - 470.00 €
Specifications
| Antigen | MRPS28 |
|---|---|
| Dilution | Western Blot 1:500-1:1000, Immunocytochemistry/ Immunofluorescence 1:20-1:100 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18694301
|
Novus Biologicals
NBP2-93219-0.02ml |
0.02 mL |
190.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18684261
|
Novus Biologicals
NBP2-93219-0.1ml |
0.1 mL |
470.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MRPS28 Polyclonal antibody specifically detects MRPS28 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| MRPS28 | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Core ESC Like Genes, Stem Cell Markers | |
| PBS (pH 7.3), 50% glycerol | |
| 28957 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:1000, Immunocytochemistry/ Immunofluorescence 1:20-1:100 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| 28S ribosomal protein S35, mitochondrial, FLJ22853, mitochondrial 28S ribosomal protein S35, mitochondrial ribosomal protein S28, MRP-S2828S ribosomal protein S28, mitochondrial, MRP-S35, MRPS35HSPC007, S28mt, S35mt | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-187 of human MRPS28 (NP_054737.1). MAALCRTRAVAAESHFLRVFLFFRPFRGVGTESGSESGSSNAKEPKTRAGGFASALERHSELLQKVEPLQKGSPKNVESFASMLRHSPLTQMGPAKDKLVIGRIFHIVENDLYIDFGGKFHCVCRRPEVDGEKYQKGTRVRLRLLDLELTSRFLGATTDTTVLEANAVLLGIQESKDSRSKEEHHEK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title